Learn More
Pyrex™ SVL™ Stirrer Heads for SVL™ Stirrer Shafts
Switch stirring heads quickly and easily. Pyrex™ SVL™ Stirrer Head for SVL™ Stirrer Shafts are constructed of durable PTFE polymer; some types are fortified with a metal core.
Brand: Pyrex™ 588-94
This item is not returnable.
View return policy
Description
- Designed for use with SVL™ stirrer shafts
- Head simply screws into the end of the shaft
- Manufactured from a PTFE based polymer with the Anchor and Helix types stiffened by a metal core
Specifications
70 mm | |
SVL™ reaction vessels | |
1 Each | |
Paddle |