Learn More
3M™ Swab-Sampler
Convenient and quick collection device for environmental sampling and testing over small surface areas.
133.56 € - 183.38 €
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
17228520
|
3M™
7000030299 |
1 mL Letheen Broth |
159.00 €
Pack of 100 |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17238520
|
3M™
7000030300 |
4 mL Letheen Broth |
133.56 €
Pack of 100 |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17248520
|
3M™
7000030301 |
4 mL Neutralizing Buffer |
169.60 €
Pack of 100 |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17258520
|
3M™
7000030302 |
5 mL Letheen Broth |
183.38 €
Pack of 100 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Letheen Broth vial is ready to use for pathogen testing of environmental samples.
- Swab reaches into corners and hard-to-reach areas for increased accuracy.
- Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
- Bottles have ample headspace for sample addition and vortexing.
- Self-contained swab and vial for increased productivity and efficiency.
Specifications
100 /Cs. | |
Swab |