Learn More
3M™ Swab-Sampler
Encompass
Convenient and quick collection device for environmental sampling and testing over small surface areas.
POA
Specifications
No. per Case 100
Product Type Swab
View More Specs
Products 3
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17203734
Encompass
View Documents
3M™
7000053817
10 mL Letheen Broth
POA
Pack of 100
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17213734
Encompass
View Documents
3M™
7000053818
10 mL Neutralizing Buffer
POA
100 sets
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17283744
Encompass
View Documents
3M™
7100010351
10 mL Buffered Peptone Water Broth
POA
Pack of 100
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Ready to use for pathogen testing of environmental samples.
  • Swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Bottles have ample headspace for sample addition and vortexing.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifications
Specifications
100
Swab
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Swab-Sampler