Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
Brand:  Thermo Scientific™ 290SG004
 View more versions of this product
Product Code. 10117414
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transfer Line Nut
Transfer Line Nut
1 Ea.
SWAP-IT GC-MS Interface System for Agilent Instruments
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.