Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
Brand:  Eppendorf™ 5804719000
 View more versions of this product
Product Code. 10054271
  • 1813.22 € / Each
Explore more special offers
Description
Description
  • Accommodates tubes and plates
  • Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
  • Rotor life time: max. 40,000 cycles
  • Rotor, buckets, caps and adapters are autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Swinging Bucket Rotor
5,000 rpm
4 hangers for 2 x 50mL Falcon-G™
4,500 x g
A-4-44
4