Learn More
SYK Mouse, anti-Human, CoraLite Plus 488, Clone: 4C4A12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66721-20UL
 View more versions of this product
Product Code. 18018765
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The SYK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SYK. This antibody recognizes human antigen. The SYK antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
SYK
4C4A12
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG2b
Western Blot
1000 μg/mL
DKFZp313N1010, FLJ25043, FLJ37489, spleen tyrosine kinase, SYK, Tyrosine protein kinase SYK
20 μL
6850
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SYK Mouse, anti-Human, CoraLite Plus 488, Clone: 4C4A12, Proteintech > Quantity: 20 μL