Learn More
Syngene™ Neutral Field Screen Set for use with IG3 and UG3 systems
GREENER_CHOICE
Contains a blue screen and a frosted screen. Syngene™ Neutral Field Screen Set is for use with IG3 and UG3 systems.
Brand:  Syngene™ GE-NFS
Product Code. 12804038
  • 37.05 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corrects for any uneven Illumination by acquiring a neutral-field reference image with the same intensity illumination as the experiment.
  • For Epi-UV lighting and transilluminators in Syngene™ Ingenius and Nugenius Systems
Specifications
Specifications
Neutral Field Screen Set
IG3 and UG3 Systems
Syngene Ingenius and Nugenius Systems
330 x 310 x 15 mm