missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene™ UV epi Light Module
Controlled GeneSys software for all fluorescence requirements. Syngene™ UV epi Light module is an optional light module with high resolution camera. Gel documentation and analysis system
1375.00 €
Product Code | Brand | Wavelength | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Type | Price | Quantity & Availability | ||||
12844068
|
Syngene™
EPI-MOD-MWSW |
302 nm, 254 nm | UV epi Light, Mid And Short Wave |
1375.00 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
12834068
|
Syngene™
EPI-MOD-SWLW |
254 nm, 365 nm | UV epi Light, Short And Long Wave |
1375.00 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
12804058
|
Syngene™
EPI-MOD-SW |
254 nm | UV epi Light, Short Wave |
1375.00 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
12824068
|
Syngene™
EPI-MOD-MWLW |
302 nm, 365 nm | UV epi Light, Mid Wave |
1375.00 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
12884048
|
Syngene™
EPI-MOD-MW |
302 nm | UV epi Light, Mid Wave |
1375.00 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
12894048
|
Syngene™
EPI-MOD-LW |
365 nm | UV epi Light, Long Wave |
1375.00 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
Description
- Odular system
- Light tight darkroom - suitable for fluorescence, upgradeable for chemiluminescence
- 16bit 3.8MP camera with auto focus, auto exposure and series capture
- Integration control - capture faint bands or features from any gel
- Neutral fielding for background correction
- Filter selection options for detection of a wide range of stains and dyes
- GeneSys software includes a database of hundreds of applications
- The system includes a motor driven zoom lens as standard and a motor driven filter wheel
- Internal lighting comes from an optional UV transilluminator or a blue light LED model to further extend the applications possible with the G:BOX F3
- Electrophoresis gels - Ethidium Bromide, Coomassie Blue, Silver stain, SYBR™ Gold, SYBR™ Green, SYBR™ Safe, GelStar™, SYPRO™ Red , SYPRO™ Ruby, SYPRO™ Orange, Fluorescein, Rhodamine Red™, Texas Red™, Pro-Q™ Diamond , Deep Purple™
- Extended fluorescence - for use with EPI RGB lighting module
- Agar plates
- Cells in flasks
- Autoradiographs
- Membranes
- Spots and slot blots
- TLC plates
- Microtitre plates
Specifications
G:BOX systems | |
Fixed |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title