missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ UV Transilluminator 2020SL
GREENER_CHOICE

Syngene™ UV Transilluminator 2020SL for illumination of any sample excited by UV light.

Brand:  Syngene™ GX-2020SL-E

Product Code. 12834028

  • 1445.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sealed to allow wet gels on surface
  • For use in Syngene™ Systems
  • 254/365nm wavelength
Specifications

Specifications

Fixed
254/364 nm
For G:Box systems
200 x 200 mm
230 V 50 Hz 230 w
Syngene 2020SL
50 Hz
UV
UV Transilluminator
230 w
335 x 310 x 100 mm
200 x 200 mm
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.