Learn More
SYNJ2BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15666-1-AP-150UL
 View more versions of this product
Product Code. 16884934
  • 410.00 € / 150µL
Explore more special offers
Description
Description
Regulates endocytosis of activin type 2 receptor kinases through the Ral/RALBP1-dependent pathway and may be involved in suppression of activin-induced signal transduction.
Specifications
Specifications
SYNJ2BP
Polyclonal
Unconjugated
SYNJ2BP
ARIP2, OMP25, synaptojanin 2 binding protein, SYNJ2BP
Rabbit
Antigen Affinity Chromatography
RUO
24071, 55333
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P57105, Q9D6K5
SYNJ2BP
SYNJ2BP Fusion Protein Ag8130
150 μL
Primary
Human, Mouse