missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Synthegel™ 3D hiPSC Matrix Kit, 10 mL
Powerful tool for the physiological formation of 3D human induced pluripotent stem cell (hiPSC) spheroids in vitro. hiPSC spheroids maintain high viability, pluripotency and maintain differentiation potential based on multiple biomarkers.
Brand: Corning™ 354787
This item is not returnable.
View return policy
Description
- Kit consists of a vial of Synthegel 3D hiPSC peptide nanofiber solution and a vial of Synthegel X-Link solution
- Requires a vial of lyophilized Synthegel 3D hiPSC Grow Mix
- Matrix rigidity is tunable by altering the peptide concentration
- Purified synthetic peptide in neutral pH
- Cells are encapsulated in physiological mimicking conditions
- Fast hydrogel formation (5 to 30 minutes)
- Sol-gel transfer reversible by shearing force
- Works with standard cell culture methodologies, growth factors and media
- Allows growth and passaging of hiPSCs in 3D format
- The hiPSC Grow Mix supplement provides a complete environment for hiPSC 3D culture
- Allows 3D embedded hiPSC cultures
Specifications
ISO | |
Store at 2 to 8°C | |
Synthegel™ 3D hiPSC Matrix Kit | |
Liquid | |
Sterile-filtered | |
3D embedded culture of hiPSC in a synthetic peptide hydrogel | |
Hydrogel | |
Ambient Temperature |
Red | |
Ambient Temperature | |
3D Cell Culture | |
Extracellular Matrix | |
Kit | |
Neutral | |
10 mL | |
Synthetic Peptide Matrix |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction