Learn More
Synthetic Sea Water for ASTM 665 / IP 135, Paragon Scientific™
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 665 / IP 135 the Standard Test Method for Rust-Preventing Characteristics of Inhibited Mineral Oil in the Presence of Water.
Brand: Paragon Scientific SSWS-5L
This item is not returnable.
View return policy
Description
- A Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D 665 / IP 135.
- Provided with a certificate which includes an estimation of uncertainty of manufacture.
- Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
- Conveniently packaged in single dose or multi dose packs.
- Cost effective and time saving
- Certificate of Manufacture & MSDS included
- 12 month shelf life
Specifications
5 L | |
ASTM 665 / IP 135 |
Product Content Correction
Product Title
Synthetic Sea Water for ASTM 665 / IP 135, Paragon Scientific™ > Quantity: 5L
Spot an opportunity for improvement?Share a Content Correction