Learn More
Synthetic Sea Water for ASTM 665 / IP 135, Paragon Scientific™
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 665 / IP 135 the Standard Test Method for Rust-Preventing Characteristics of Inhibited Mineral Oil in the Presence of Water.
Brand:  Paragon Scientific SSWS-500
 View more versions of this product
Product Code. 16090420
  • 246.00 € / 500mL
Explore more special offers
Description
Description
  • A Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D 665 / IP 135.
  • Provided with a certificate which includes an estimation of uncertainty of manufacture.
  • Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
  • Conveniently packaged in single dose or multi dose packs.
  • Cost effective and time saving
  • Certificate of Manufacture & MSDS included
  • 12 month shelf life
Specifications
Specifications
500 mL
ASTM 665 / IP 135
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Synthetic Sea Water for ASTM 665 / IP 135, Paragon Scientific™ > Quantity: 500mL