missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ SYPRO™ Protein Gel Stains

Liquid format protein gel stains

257.00 € - 836.00 €

Specifications

Product Type Protein Stain
Form Liquid
Storage Requirements 4°C
View More Specs

Products 2
Product Code Brand Color Quantity Product Type Price Quantity & Availability  
Product Code Brand Color Quantity Product Type Price Quantity & Availability  
11520777
View Documents
Lonza™
50562
Ruby 1 L Protein Stain
836.00 €
Each
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
11540777
View Documents
Lonza™
50564
Ruby 200 mL Protein Stain
257.00 €
Each
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
Description

Description

SYPRO Ruby Protein Gel Stains

  • Ideal for 2-D gel analysis Sensitivity rivals silver stain Detects nanogram levels of protein Offers a broad linear quantitation range and consistent gel-to-gel staining for accurate protein expression comparisons
SYPRO Tangerine Protein Gel Stain
  • Provides high sensitivity for detection of proteins in SDS-polyacrylamide gels
  • Does not require the use of acids or organic solvents
  • Suitable for use with saline solutions Ideal for use when eluting active protein complexes from gels because the mild staining conditions are unlikely to affect protein activity
SYPRO Red Protein Gel Stain
  • Detects as little as 4 to 8ng of protein per band or proteins as small as 6.5kDa Provides linear protein quantitation up to three orders of magnitude, from ng to μg per band
  • Offers complete staining in less than one hour—no fixation or destaining required
  • Compatible with many types of detection systems Recommended for instruments that employ frequency-doubled Nd:YAG or green or red He-Ne lasers

Specifications

Specifications

Protein Stain
Liquid
4°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 6 to 12 Months

SDS
Documents

Documents

Product Certifications