Learn More
Socorex™ Dosys™ All-glass Syringes
Resistant borosilicate glass syringes can be sterilized and reused. Socorex™ Dosys™ All-glass Syringes provide a low-cost alternative to disposable plastic syringes and are easy to disassemble for cleaning.
Brand:  Socorex™ 155.0502
 View more versions of this product
Product Code. 10477072
  • 32.20 € / Pack of 3
Explore more special offers
Description
Description
  • Borosilicate glass construction.
  • Superior chemical, heat and shock resistance.
  • Low-cost alternative to disposable plastic syringes.
  • Precision-machined plunger and barrel.
  • Excellent fitting and liquid tightness (not gas tight).
  • Permanent, high visibility graduations.
  • Glass Luer and metal Luer lock nozzles available.
  • Can be sterilized up to 160°C / 320°F.
  • Very easy to disassemble and clean.
Specifications
Specifications
Autoclavable
Syringe
3/Pk.
Luer Lock
Glass, Glass (Barrel), Glass (Plunger), Metal (Tip)
0.1 mL
0.5 to 2 mL
Yes