Learn More
Sartorius Syringe Holder/Support for Sterisart™ 16469
The pre-filled Syringe Holder/Support for Sterisart™ 16469 facilitates dilution of the contents of pre-filled syringes and flushing the surface of a syringe needle, during sterility testing.
Brand:  Sartorius 1ZA---0024
Product Code. 15972600
  • 9285.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Syringe holder and rinsing fluid flow-rate controller/restrictor
  • Compatible with:
    • Sterisart™ 16469--------GBD
    • Sterisart™ 16469--------GSD
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterisart System 16469
Syringe Holder