Learn More
Hamilton™ Custom Cemented Needle Syringes
The gas-tight version of the original 700 series syringes.
Brand:  Hamilton™ 202134
 View more versions of this product
Product Code. 10281973
  • 116.00 € / Each
Explore more special offers
Description
Description
  • Ideal for dispensing volumes from 1 up to 500 μL
  • A variety of volumes, lengths, and gauges available
Specifications
Specifications
Not Autoclavable
1725
250 μL
Syringe Needle
1 Ea.