Learn More
Hamilton™ Custom Cemented Needle Syringes
The gas-tight version of the original 700 series syringes.
Brand:  Hamilton™ 80208
 View more versions of this product
Product Code. 10491245
  • 181.00 € / Each
Explore more special offers
Description
Description
  • Ideal for dispensing volumes from 1 up to 500 μL
  • A variety of volumes, lengths, and gauges available
Specifications
Specifications
Not Autoclavable
0.729 mm
Gastight
50°C
10°C
25 μL
Custom
Custom
7.75 mm
68.9 bar
1702
1 Ea.
Custom Needle
Cemented