missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Syringes for Thermo Scientific™ HPLC Instruments

Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.

Brand:  Thermo Scientific™ 365JLT41

162.70 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10341205

  • 197.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.

  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications

Specifications

22 G
50 mm
Reusable
LCQ HPLC
2 in. Removable Needle
HPLC Autosampler Syringe
500 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.