missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ Green Fluorescent Nucleic Acid Stains

SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria.

Brand:  Invitrogen™ S7572

503.12 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10646543

  • 662.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria. Because the dye characteristics and staining pattern of the SYTO dyes may vary, we offer 10 different SYTO Green Fluorescent Nucleic Acid Stains available individually along with the SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 (containing SYTO 11–14, 16, 21, and 24) to enable researchers to find the most appropriate green-fluorescent SYTO stain for their system . We also provide the SYTOX Green Nucleic Acid Stain (Cat. No. S7020 and R37109), which is a cell-impermeant green-fluorescent stain used for staining dead cells or performing nuclear counterstaining in fixed cells.

The SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 contains our collection of cell-permeant, green-fluorescent SYTO nucleic acid stains. Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL each of SYTO 11–14, 16, 21, and 24 dyes.

Any physiological buffer that is pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.
 

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
SYTO 11 DNA: 508 nm/RNA: 510 nm
SYTO 14 DNA: 517 nm/RNA: 521 nm
SYTO 16 DNA: 488 nm/RNA: 494 nm
SYTO 21 DNA: 494 nm
SYTO 24 DNA: 490 nm
Fluorescent Labeling, Live Cell Imaging, Microarray
1 kit
Fluorescence
SYTO 11 DNA: 527 nm/RNA: 530 nm
SYTO 14 DNA: 549 nm/RNA: 547 nm
SYTO 16 DNA: 518 nm/RNA: 525 nm
SYTO 21 DNA 517 nm
SYTO 24 DNA: 515 nm
Room Temperature
Stains Dimer Sampler Kit
The kit contains 50 μL each of SYTO 11–14, 16, 21, and 24 dyes.

Store in freezer at –5 to –30°C and protect from light.
Cell-Permeant
Fluorescence Microscope, High Content Imager, Flow Cytometer
50 μL
SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1: SYTO dyes 11, 14, 16, 21, 24 (50 μL each)
SYTO™
Fluorescent Dye
Nucleic Acids
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.