missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ SYTOX™ Blue Dead Cell Stain, for flow cytometry

SYTOX™ Blue dead cell stain is a simple and quantitative single-step dead-cell indicator for use with violet laser equipped flow cytometers.

Brand:  Molecular Probes™ S34857

558.60 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 10297242

  • 588.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A simple and quantitative single-step dead-cell indicator for use with violet laser equipped flow cytometers
  • SYTOX™ Blue dead cell stain is a high-affinity nucleic acid stain that easily penetrates cells with compromised plasma membranes but will not cross uncompromised cell membranes
  • After brief incubation with SYTOX™ Blue stain, the nucleic acids of dead cells fluoresce bright blue when excited with 405 nm violet laser light
  • The violet-excited fluorescence emission permits clear discrimination from probes excited by most other laser lines, facilitating the development of multicolor assays with minimal spectral overlap between signals
  • Supplied as a 1 mM solution in dimethylsulfoxide (DMSO)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mM
Fluorescence
Flow Cytometer
SYTOX™ Blue
Solution
SYTOX™
Room Temperature
Contains 1 vial of SYTOX™ blue dead cell stain (1 mM solution in DMSO).

Store in freezer (-5 to -30°C), desiccated and protect from light.
Viability Assay
Dead Cell Stain
444/480
Tube(s)
1 mL
Nucleus, Nucleic Acids
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.