missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTOX™ Green Ready Flow™ Reagent

SYTOX™ Green Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays.

Brand:  Invitrogen™ R37168

241.87 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 15983460

  • 361.00 € / Pack of 120

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays
  • It is excitable at 488 nm with an emission profile similar to FITC, and has been tested in multiple cell lines
  • Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze
  • As a cell-impermeant dye, SYTOX Green enters the cell upon loss of membrane integrity and binds to DNA, thereby acting as a counterstain that can be analyzed when excited
  • SYTOX Green reagent can be used in both mammalian and bacterial cells
  • It is often included in multiplex assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Impermeant
Includes 2 dropper bottles, store at room temperature.
Fluorescence
Flow Cytometer
SYTOX™ Green Ready Flow™
504 nm
Bottle(s)
120 Assays
Green
SYTOX™ Green Ready Flow™ Reagent
Viability Assay
Reagent
523 nm
Solution
Ready Flow™, Sytox™
DMSO (Dimethylsulfoxide)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2980694 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog NumberR37168
Lot Number2800329 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog NumberR37168
Lot Number2714535 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberR37168
Lot Number2647609 Certificate TypeCertificate of Analysis Date07/06/2023 Catalog NumberR37168
Lot Number2550779 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog NumberR37168
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SYTOX™ Green Ready Flow™ Reagent > 120 assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.