missing translation for 'onlineSavingsMsg'
Learn More

T 0901317, Tocris Bioscience™

Product Code. 18094502
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent liver X receptor (LXR) agonist; also CAR inverse agonist

Potent, high affinity liver X receptor (LXR) agonist (EC50 ∽ 50 nM, Kd values are 7 and 22 nM for LXR-α and LXR-β respectively). Upregulates expression of the ABCA1 gene associated with cholesterol efflux regulation and HDL metabolism. Decreases amyloid-β production in primary neurons in vitro. Displays an EC50 of ∽ 5 μM for activation of bile acid farnesoid X receptors (FXRs); 10-fold more potent than natural FXR ligand chenodeoxycholic acid. Also exhibits inverse agonist activity at constitutive androstane receptors (CAR).

Chemical Identifiers

CAS 293754-55-9
Molecular Formula C17H12F9NO3S
Molecular Weight (g/mol) 481.333
InChI Key SGIWFELWJPNFDH-UHFFFAOYSA-N
Synonym n-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl-n-2,2,2-trifluoroethyl benzenesulfonamide, chembl62136, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl benzenesulfonamide, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethylethyl phenyl sulfonamide, 1pqc, d0b5fc, n-4-1,1,1,3,3,3-hexafluoro-2-hydroxy-propan-2-yl phenyl-n-2,2,2-trifluoroethyl benzenesulfonamide, t, benzenesulfonamide, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl
PubChem CID 447912
ChEBI CHEBI:39976
IUPAC Name N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N(CC(F)(F)F)C2=CC=C(C=C2)C(C(F)(F)F)(C(F)(F)F)O

Specifications

Quantity 10 mg
Formula Weight 481.33
Percent Purity >98%
Chemical Name or Material T 0901317
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.