missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ T-1270 Fixed Angle Titanium Rotor
Effectively perform a wide range of applications with the T-1270 Rotor, which offers 12-place capacity.
Brand: Thermo Scientific™ 08259
20848.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Effectively perform a wide range of applications with the Thermo Scientific™ T-1270 Fixed Angle Rotor, which offers 12-place capacity.
Recommended for:DNA isolation, serum flotation.
Specifications
T-1270 Fixed Angle Titanium Rotor | |
Fixed Angle Rotor | |
448,811 x g | |
82 mm | |
36.3 | |
16 mm | |
Thermo Scientific™ Sorvall™ WX ultraCentrifuges | |
335,513 x g |
Titanium | |
12 x 12.5 mL | |
70,000 rpm | |
40.6 mm | |
80 mm | |
13 lb. (5.9 kg) | |
24° | |
222,216 x g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction