Learn More
GVS T200 Respirator with Full Hood and Internal Face Seal
The T200 respirator provides great protection for those in healthcare, EMS, and other types of medical enviroments. Very lightweight and comfortable to help protect the user better. It provides a wide viewing area to make your work easier.
Brand:  GVS 17-200-32-CE
Product Code. 18489655
  • 41.30 € / Each
Explore more special offers
Description
Description
The T200 respirator provides great protection for those in healthcare, EMS, and other types of medical enviroments.
  • Very lightweight and comfortable to help protect the user better. It provides a wide viewing area to make your work easier.
  • Multi adjustment to fit a wide range of users.
  • Certified to EN 12941 1998 +A2:2008 and EN 166 2001 1 F 3.
  • Includes: Full hood with internal face seal, air duct and head harness assembly, 04-831 PX5™ breathing tube
Specifications
Specifications
White
PAPR System with Hood and Head Harness
One Size