Learn More
Thermo Scientific™ T4 DNA Ligase (5 U/μL)
T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.
47.79 € - 322.00 €
Specifications
Concentration 5 U/μL
Enzyme DNA Ligase
Compatible Buffer 10X T4 DNA Ligase Buffer
Product Type T4 DNA Ligase
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10723941
View Documents
Thermo Scientific™
EL0011
1,000 units
82.75 €
1000 units
10621441
View Documents
Thermo Scientific™
EL0012
5 x 1,000 units
322.00 €
5000 units
10339509
View Documents
Thermo Scientific™
EL0014
200 units
47.79 €
200 units
Description
Show More
Specifications
Specifications
5 U/μL
DNA Ligase
10X T4 DNA Ligase Buffer
T4 DNA Ligase
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ T4 DNA Ligase (5 U/μL)