Learn More
Sampling Systems™ Tablet Counting Triangle
Sampling Systems™ Tablet Counting Triangle is ideal for counting tablets in hygienic and pharmaceutical applications.
Brand:  Sampling Systems™ A356-275
Product Code. 15605859
  • 91.50 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quick and easy way to count round tablets
  • Suitable for all sizes of round tablets
  • 316L Stainless Steel
  • Satin finish
  • Can be easily used by right or left handed operators
  • Etched with a guide to show the number of tablets in the tray
Specifications
Specifications
12 mm
275 mm