missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TABS, 99%

113.57 € - 323.61 €

Chemical Identifiers

CAS 54960-65-5
Synonym N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10159959
SDS View Documents
Thermo Scientific Chemicals
428080250
25 g Glass bottle
113.57 €
25g
Please to purchase this item. Need a web account? Register with us today!
10311800
SDS View Documents
Thermo Scientific Chemicals
428081000
100 g Glass bottle
323.61 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

54960-65-5
N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid

Specifications

257.301
N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid
≥98.5% (on anhydrous basis)
54960-65-5
Crystalline
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TABS

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals TABS, 99%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.