missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURUS™ 60-Layer Biohybrid™ Contamination Control Mats
PURUS™ Biohybrid™ Mats are the first ever contamination control products that contain renewable plant-based materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels
Brand: PURUS™ PS 3636 64 W
231.80 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Earth Friendly containing GMO free, renewable plant based Cardia Biohybrid™ material that reduces CO2 emissions
- Strong & Effective delivering reliable and high-end performance
- Consistent adhesive coverage for maximum performance of each layer Can be recycled in conventional plastic recycling streams
- Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Specifications
Manufactured in an ISO 9001 certified facility | |
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications | |
Regular Tack | |
Film: 33 μm Adhesive: 8 μm |
914 x 914 mm | |
60 layers per mat, 4 mats per case | |
Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | |
White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction