Learn More
IBA Lifesciences TACS Column Adapter
Adapter for Strep-Tactin™ TACS Agarose Columns that fits onto standard tubes
11.30 €
Product Code | Brand | Column Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Type | Price | Quantity & Availability | |||||
17277590
|
IBA Lifesciences
6-3333-001 |
0.3 mL |
11.30 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
15819794
|
IBA Lifesciences
6-6331-001 |
1 mL |
11.30 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
Description
- The adapters allow fitting of Strep-Tactin™ TACS Agarose columns onto standard tubes.
- Column adapters are available in two sizes: For 3 ml columns (containing 0.3 ml Strep-Tactin™ TACS Agarose) to fit them onto 15 ml tubes and for 10 ml columns (containing 1 ml Strep-Tactin™ TACS Agarose) to fit them onto 50 ml tubes.