Learn More
Sartorius Tacta™ Mechanical Pipettes
BIOPHARMAGREENER_CHOICE
Product Code. 15596304 Shop All Sartorius Products
Click to view available options
Number of Channels:
1
8
12
Volume (Metric):
0.1 to 3 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 10,000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 100 μL
500 to 5000 μL
Color:
Blue
Gray
Green
Orange
Red
Yellow
Unit Size:
Each
Product Code. 15596304
Brand: Sartorius LH729050
Only null left
Add to basket
Edge
Add to basket
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Optifit 200 μL, 350 μL, SafetySpace Filter Tips 120 μL
Adjustable Tip Spacing Yes
Autoclavable Autoclavable
Color Yellow
Disposable Reusable
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Product Line Tacta
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 10 to 100 μL
Color Code Yellow