missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
68.40 € - 158.00 €
Specifications
Chemical Name or Material | TAE Buffer |
---|---|
Concentration | 10X |
Physical Form | Solution |
Grade | Molecular Biology, Ultrapure |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15885418
|
Thermo Scientific
J75904.K2 |
1 L | Nalgene bottle |
68.40 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15895418
|
Thermo Scientific
J75904.K8 |
5 L | Upright carton |
158.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Specifications
TAE Buffer | |
10X | |
Solution | |
Molecular Biology, Ultrapure |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title