missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tali™ Cellular Analysis Slides

The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer.

295.00 € - 2730.00 €

Specifications

Type Cellular Analysis Slide
Product Line Tali
For Use With (Equipment) Tali™ Image-Based Cytometer
Detection Method Fluorescence
Shipping Condition Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12083805
View Documents Promotion Details
Invitrogen™
T10794
50 Slides
295.00 €
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
12209319
View Documents Promotion Details
Invitrogen™
T10795
500 Slides
2730.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer. Each slide contains two separate, enclosed analysis chambers designed to accommodate 25 μL of sample per chamber. Each box contains 50 slides.

Ready to Use for Your Convenience

Each Tali™ Cellular Analysis slide is pre-sterilized and individually packaged. Since the Tali™ Image-Based Cytometer is a fluorescent cellular analysis instrument the slides are composed of low fluorescent plastic. The slides are disposable; after use the slides can be disposed of with you regular biohazard waste.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Cellular Analysis Slide
Tali™ Image-Based Cytometer
Room Temperature
Tali
Fluorescence
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.