Learn More
Kartell™ Tall Form Graduated Measuring Cylinders
Accurately measures the volume of a liquid. Kartell™ Tall Form Graduated Measuring Cylinders use horizontal marked lines to assist in measuring out various volumes of a liquid.
41.75 € - 229.00 €
Specifications
Class B
Autoclavable Autoclavable
View More Specs
Products 7
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
15845577
View Documents
Kartell™
0257300
10 mL
229.00 €
Pack of 30
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
12593267
View Documents
Kartell™
0257400
20 mL
193.00 €
Pack of 12
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
15815577
View Documents
Kartell™
0257000
10 mL 2 mL
41.75 €
Pack of 10
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
15825577
View Documents
Kartell™
0257100
5 mL
177.00 €
Pack of 30
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
12655686
View Documents
Kartell™
0257700
200 mL
135.00 €
Pack of 2
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
15865577
View Documents
Kartell™
0257600
100 mL
173.00 €
Pack of 6
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
15835577
View Documents
Kartell™
0257200
50 mL 10 mL
201.00 €
Pack of 30
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
Description
Description
  • Tall form
  • Circular base
  • Good chemical resistance
  • Permanent graduations moulded into cylinders
  • Can withstand temperatures up to 170°C
  • Suitable for applications with food
  • No wetting and no meniscus
Specifications
Specifications
B
Autoclavable