Learn More
Thermo Scientific Chemicals Tamoxifen citrate, 98%
260.00 € - 897.00 €
Chemical Identifiers
CAS | 54965-24-1 |
---|---|
Molecular Formula | C32H37NO8 |
Molecular Weight (g/mol) | 563.65 |
MDL Number | MFCD00058321 |
InChI Key | FQZYTYWMLGAPFJ-OQKDUQJOSA-N |
Synonym | tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen |
PubChem CID | 2733525 |
ChEBI | CHEBI:9397 |
IUPAC Name | 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
SMILES | OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10143141
|
Thermo Scientific Chemicals
329320010 |
1 g |
260.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10155633
|
Thermo Scientific Chemicals
329320050 |
5 g |
897.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54965-24-1 | |
563.65 | |
FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
2733525 | |
2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
C32H37NO8 | |
MFCD00058321 | |
tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen | |
CHEBI:9397 | |
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 |
Specifications
54965-24-1 | |
White | |
98% | |
C32H37NO8 | |
15, 9180 | |
Solubility in water: slightly soluble in water. Other solubilities: soluble in methanol, ethanol, acetone | |
OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 | |
563.65 | |
CHEBI:9397 | |
98% | |
Tamoxifen citrate, 98% |
140.0°C to 144.0°C | |
Authentic | |
Glass bottle | |
MFCD00058321 | |
tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen | |
FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid | |
2733525 | |
563.65 | |
Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause cancer.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 259-415-2
missing translation for 'rtecsNumber' : KH2387000
- Tamoxifen citrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
Hazard Statement
- H350-May cause cancer.
- H302-Harmful if swallowed.
- H360-May damage fertility or the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only