Learn More
Brady™ Tamper-evident Thermal Transfer Vinyl Labels
Printable, tamper-evident vinyl labels for Thermal Transfer Printers.
Brand:  Brady™ 030242
 View more versions of this product
Product Code. 11573804
  • 270.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to fracture easily in order to show signs of product tampering and prevent one-piece label removal
  • Use caution when removing from liner, as material is fragile
Specifications
Specifications
Not Autoclavable
White
Vinyl (Tamper-resistant)
Non-sterile
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
4
Rectangle
0.066 mm
16.5 mm
Permanent Acrylic
Satin
Thermal Transfer
Circuit Board Labeling, Security Identification
17.8 mm
10,000 Labels
-40°C to +80°C
7.6 mm
5.1 mm