missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ TAMRA, SE; 5-(and-6)-Carboxytetramethylrhodamine, Succinimidyl Ester (5(6)-TAMRA, SE), mixed isomers
TAMRA, SE; 5-(and-6)-Carboxytetramethylrhodamine, Succinimidyl Ester (5(6)-TAMRA, SE), mixed isomers
Brand: Molecular Probes™ C1171
397.32 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Tetramethylrhodamine (TMR) is an important fluorophore for preparing protein conjugates. Under the name TAMRA, the carboxylic acid of TMR has also achieved prominence as a dye for oligonucleotide labeling and automated DNA sequencing applications. 5-(and-6)-carboxytetramethylrhodamine, succinimidyl ester is the amine-reactive, mixed isomer form of TAMRA. Single isomer forms of 5-TAMRA SE (Cat no. C-2211) and 6-TAMRA SE (Cat. no. C-6123) are also available.Specifications
25 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
TAMRA™ Isomers, TMR (Tetramethylrhodamine) |
TAMRA SE | |
Amine | |
Active Ester, Succinimidyl Ester | |
Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.