Learn More
Thermo Scientific™ Tango™ ADRA2A-bla U2OS Cells
The Tango™ ADRA2A-bla U2OS cells contain the human Adrenergic, Alpha-2A-, Receptor (ADRA2A) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™ K1604
Description
The Tango™ ADRA2A-bla U2OS cells contain the human Adrenergic, Alpha-2A-, Receptor (ADRA2A) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ ADRA2A-bla U2OS cells cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ ADRA2A-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
Specifications
NM_000681.2 | |
Dividing Cells | |
Tail from AVPR2 GPCR | |
GPCRs | |
ADRA2A-bla U2OS cells, ADRA2A-bla U2OS Assay Protocol, Certificate of Analysis | |
Tango™ | |
BLA (Beta-Lactamase) | |
Dividing Cells | |
Tango™ |
Cell-based beta-arrestin recruitment | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ ADRA2A-bla U2OS cells • Tango™ ADRA2A-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ ADRA2A-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
ADRA2A | |
Single Cell Clone Origin | |
2 x 106 Cells | |
ADRA2A | |
U2OS | |
GPCR |
For Research Use Only. Not for use in diagnostic procedures.