Learn More
Thermo Scientific™ Tango™ CMKLR1-bla U2OS Cells
The Tango™ CMKLR1-bla U2OS cells contain the human Chemokine like receptor 1 (CMKLR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1527
Product Code. 10701434
  • 12780.00 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The Tango™ CMKLR1-bla U2OS cells contain the human Chemokine like receptor 1 (CMKLR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_004072.1
Dividing Cells
Native Tail
Fluorescence
CMKLR1
Single Cell Clone Origin
2 x 106 Cells
U2OS
CMKLR1
U2OS
GPCR
Cell-based beta-arrestin recruitment
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ CMKLR1-bla U2OS cells
• Tango™ CMKLR1-bla U2OS Assay Protocol
• Certificate of Analysis
The Tango™ CMKLR1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Tango™ CMKLR1-bla U2OS Cells, 2 x 106 Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
Tango™ CMKLR1-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ CMKLR1-bla U2OS cells, Tango™ CMKLR1-bla U2OS assay protocol, certificate of analysis
Tango™
BLA (Beta-Lactamase)
Room Temperature
U2OS Cell
Tango™