missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tango™ CXCR3-bla U2OS Cells

The Tango™ CXCR3-bla U2OS cells contain the human chemokine (C-X-C motif) receptor 3 (CXCR3) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

Brand:  Thermo Scientific™ K1829

Product Code. 10471215

  • 12220.00 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The Tango™ CXCR3-bla U2OS cells are functionally validated for Z' and EC50 concentrations of I-TAC (CXCL11). In addition, Tango™ CXCR3-bla U2OS cells have been tested for assay performance under variable conditions. This data are found in the Validation & Assay Performance Summary.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_001504
Dividing Cells
Native Tail
FRET
G-Protein Coupled Receptor (GPCR)
GeneBLAzer™, Tango™
End Point
U2OS Cell
Tango™
Cell-based beta-arrestin recruitment
Contents:
K1829 Tango™ CXCR3-bla U2OS cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• Tango™ CXCR3-bla U2OS cells
• Tango™ CXCR3-bla U2OS Assay Protocol
• Certificate of Analysis

Storage:
The Tango™ CXCR3-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Fluorescence
CXCR3
Single Cell Clone Origin
8 x 106 Cells
CXCR3
U2OS
GPCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.