Learn More
Thermo Scientific™ Tango™ GPR119-bla U2OS Cells
The Tango™ GPR119-bla U2OS cells contain the human G-protein Coupled Receptor 119 (GPR119) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1777
Product Code. 10522304
  • 13210.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Tango™ GPR119-bla U2OS cells contain the human G-protein Coupled Receptor 119 (GPR119) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ GPR119-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ GPR119-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_178471.1
Dividing Cells
Native Tail
GPCRs
GPR119-bla U2OS cells, GPR119-bla U2OS Assay Protocol, Certificate of Analysis
GeneBLAzer™
8 x 106 Cells
BLA (Beta-Lactamase)
Dividing Cells
Tango™
Cell-based beta-arrestin recruitment
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ GPR119-bla U2OS cells
• Tango™ GPR119-bla U2OS Assay Protocol
• Certificate of Analysis
The Tango™ GPR119-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Fluorescence
GPR119
Single Cell Clone Origin
GPCR (G-Protein Coupled Receptors)
End Point
GPR119
U2OS
GPCR