Learn More
Thermo Scientific™ Tango™ MTNR1A-bla U2OS Cells
The Tango™ MTNR1A-bla U2OS cells contain the human Melatonin Receptor 1A (MTNR1A) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™ K1669
Description
This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ MTNR1A-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ MTNR1A-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
NM_005958.3 | |
Dividing Cells | |
Native Tail | |
MTNR1A | |
GeneBLAzer™ | |
8 x 106 cells | |
BLA (Beta-Lactamase) | |
U2OS | |
GPCR |
Cell-based beta-arrestin recruitment | |
K1669 Tango™ MTNR1A-bla U2OS cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: • Tango™ MTNR1A-bla U2OS cells • Tango™ MTNR1A-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ MTNR1A-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
Single Cell Clone Origin | |
GPCR (G-Protein Coupled Receptors) | |
End Point | |
MTNR1A | |
Tango™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.