Learn More
Thermo Scientific™ Tango™ OPRM1-bla U2OS DA Assay Kit
The Tango™ OPRM1-bla U2OS DA (Division Arrested) cells contain the human Opioid Receptor Mu 1 (OPRM1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1599
Product Code. 10521824
  • 840.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Tango™ OPRM1-bla U2OS DA (Division Arrested) cells contain the human Opioid Receptor Mu 1 (OPRM1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ OPRM1-bla U2OS DA cells have been functionally validated for Z' factor and EC50 concentrations of DAMGO. In addition, Tango™ OPRM1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_000914
Division-Arrested Cells
Tail from AVPR2 GPCR
Fluorescence
OPRM1
Single Cell Clone Origin
1 Plate
U2OS
Division-Arrested Cells Assay Kit
Tango™
Cell-based beta-arrestin recruitment
Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol).
Includes:
• Tango™ OPRM1-bla U2OS DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D
• Tango™ OPRM1-bla U2OS Assay Protocol
• LiveBLAzer™-FRET B/G Loading Protocol
• Certificate of Analysis
The Tango™ OPRM1-bla U2OS DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Tango™ OPRM1-bla U2OS DA Assay Kit, 1 Plate, ∽8,000,000 cells/vial Quantity of Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol). Includes: Tango™ OPRM1-bla U2OS DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg, Solution D, Tango™ OPRM1-bla U2OS Assay Protocol, LiveBLAzer™-FRET B/G Loading Protocol, Certificate of Analysis
Tango™
BLA (Beta-Lactamase)
OPRM1
U2OS
GPCR