Learn More
Tanshinone I, MedChemExpress
MedChemExpress Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).
Brand: Medchem Express HY-N0134
This item is not returnable.
View return policy
Description
Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).Specifications
Tanshinone I | |
Dark Brown | |
C18H12O3 | |
Tanshinone A | |
O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC=C4C)=O | |
276.29 | |
Research | |
COVID-19-immunoregulation |
568-73-0 | |
Research | |
50 mg | |
DMSO : 2 mg/mL (7.24 mM; Need ultrasonic) | |
276.29 | |
98.0% | |
Solid |
Research purposes only