Learn More
Tanshinone IIA, MedChemExpress
MedChemExpress Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Brand:  Medchem Express HY-N0135
Encompass_Preferred
 View more versions of this product
Product Code. 16444767
  • 306.45 € / 50mg
Explore more special offers
Description
Description
Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Specifications
Specifications
Tanshinone IIA
Red
C19H18O3
Dan Shen ketone
O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC=C4C)=O
294.34
Research
COVID-19-immunoregulation
568-72-9
Research
50 mg
DMSO : 100 mg/mL (339.74 mM; Need ultrasonic)
294.34
99.17%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Tanshinone IIA, MedChemExpress