Learn More
Tanshinone IIA, MedChemExpress
MedChemExpress Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Brand:  Medchem Express HY-N0135
Encompass_Preferred
 View more versions of this product
Product Code. 16493836
  • 167.40 € / 25mg
Explore more special offers
Description
Description
Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Specifications
Specifications
Tanshinone IIA
Red
C19H18O3
Dan Shen ketone
O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC=C4C)=O
294.34
Research
COVID-19-immunoregulation
568-72-9
Research
25 mg
DMSO : 100 mg/mL (339.74 mM; Need ultrasonic)
294.34
99.17%
Solid