missing translation for 'onlineSavingsMsg'
Learn More
Tantalum carbide, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11306518

Tantalum carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11306518
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11306518

Brand: Thermo Scientific Alfa Aesar 012144.18

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tantalum carbides is used in  tungsten carbide. It is also used as a coating for steel moulds in the injection moulding of aluminum alloys. While providing a hard, wear resistant surface, it also provides a low friction mould surface. It is also used in the production of sharp instruments with extreme mechanical resistance and hardness, tool bits for cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-06-3
Molecular Formula CTa
Molecular Weight (g/mol) 192.96
MDL Number MFCD00011255
InChI Key DUMHRFXBHXIRTD-UHFFFAOYSA-N
IUPAC Name methanidylidynetantalumylium
SMILES [C-]#[Ta+]

Specifications

Boiling Point 5500°C
Melting Point 3880°C
Color Black to Gray
Physical Form Powder
Quantity 50 g
UN Number UN3178
Solubility Information Soluble in hydrofluoric acid and nitric acid. Partially soluble in water.
Formula Weight 192.96
Percent Purity 99.5%
Odor Odorless
Density 13.9 g/mL
Assay (metals basis)
Chemical Name or Material Tantalum carbide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum carbide, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.