missing translation for 'onlineSavingsMsg'
Learn More

Tantalum disilicide, Thermo Scientific Chemicals

66.33 € - 286.43 €

Chemical Identifiers

CAS 12039-79-1
Molecular Formula Si2Ta
Molecular Weight (g/mol) 237.118
MDL Number MFCD00049564
InChI Key MANYRMJQFFSZKJ-UHFFFAOYSA-N
Synonym tantalum disilicide, tantalum silicide tasi2, tantalum silicide, tasi2
PubChem CID 6336888
IUPAC Name bis($l^{2}-silanylidene)tantalum
SMILES [Si]=[Ta]=[Si]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15432087
SDS View Documents
Thermo Scientific Alfa Aesar
044965.09
10 g
66.33 €
10g
Please to purchase this item. Need a web account? Register with us today!
15442087
SDS View Documents
Thermo Scientific Alfa Aesar
044965.18
50 g
286.43 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tantalum disilicide is used in the preparation of ceramics, which finds application in the thermal protection systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12039-79-1
237.118
MANYRMJQFFSZKJ-UHFFFAOYSA-N
6336888
[Si]=[Ta]=[Si]
Si2Ta
MFCD00049564
tantalum disilicide, tantalum silicide tasi2, tantalum silicide, tasi2
bis($l^{2}-silanylidene)tantalum

Specifications

2200°C
Powder
MFCD00049564
Insoluble in water.
[Si]=[Ta]=[Si]
237.118
237.12
12039-79-1
Si2Ta
tantalum disilicide, tantalum silicide tasi2, tantalum silicide, tasi2
MANYRMJQFFSZKJ-UHFFFAOYSA-N
bis($l^{2}-silanylidene)tantalum
6336888
Tantalum disilicide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-902-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.