missing translation for 'onlineSavingsMsg'
Learn More
Tantalum nitride, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11312188

Tantalum nitride, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11312188
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11312188

Brand: Thermo Scientific Alfa Aesar 013093.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tantalum nitride is used to create barrier or glue layers between copper, or other conductive metals, and dielectric insulator films such as thermal oxides. These films are deposited on top of silicon wafers during the manufacture of integrated circuits, to create thin film surface mount resistors and has other electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12033-62-4
Molecular Formula NTa
Molecular Weight (g/mol) 194.955
MDL Number MFCD00049568
InChI Key MZLGASXMSKOWSE-UHFFFAOYSA-N
Synonym tantalum nitride, tantalum nitride tan, tantalum mononitride, tantalum nitride trace metals basis 10g
PubChem CID 82832
IUPAC Name azanylidynetantalum
SMILES N#[Ta]

Specifications

Melting Point 3360°C
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 194.95
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Tantalum nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum nitride, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.