missing translation for 'onlineSavingsMsg'
Learn More

Tantalum nitride, 99.5% (metals basis), Thermo Scientific Chemicals

76.48 € - 296.48 €

Chemical Identifiers

CAS 12033-62-4
Molecular Formula NTa
Molecular Weight (g/mol) 194.955
MDL Number MFCD00049568
InChI Key MZLGASXMSKOWSE-UHFFFAOYSA-N
Synonym tantalum nitride, tantalum nitride tan, tantalum mononitride, tantalum nitride trace metals basis 10g
PubChem CID 82832
IUPAC Name azanylidynetantalum
SMILES N#[Ta]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302188
View Documents
Thermo Scientific Alfa Aesar
013093.09
10 g
76.48 €
10g
Please to purchase this item. Need a web account? Register with us today!
11312188
View Documents
Thermo Scientific Alfa Aesar
013093.18
50 g
296.48 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tantalum nitride is used to create barrier or glue layers between copper, or other conductive metals, and dielectric insulator films such as thermal oxides. These films are deposited on top of silicon wafers during the manufacture of integrated circuits, to create thin film surface mount resistors and has other electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12033-62-4
194.955
MZLGASXMSKOWSE-UHFFFAOYSA-N
82832
N#[Ta]
NTa
MFCD00049568
tantalum nitride, tantalum nitride tan, tantalum mononitride, tantalum nitride trace metals basis 10g
azanylidynetantalum

Specifications

3360°C
Powder
MFCD00049568
Insoluble in water.
N#[Ta]
194.955
194.95
Odorless
Tantalum nitride
12033-62-4
NTa
tantalum nitride, tantalum nitride tan, tantalum mononitride, tantalum nitride trace metals basis 10g
MZLGASXMSKOWSE-UHFFFAOYSA-N
azanylidynetantalum
82832
99.5%
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-788-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.